Skip to Content
Merck
All Photos(1)

Key Documents

H4506

Sigma-Aldrich

1-Heptanethiol

98%

Synonym(s):

Heptyl mercaptan, Mercaptan C7

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6SH
CAS Number:
Molecular Weight:
132.27
Beilstein:
1731687
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.4520 (lit.)

bp

173-176 °C/765 mmHg (lit.)

density

0.844 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCS

InChI

1S/C7H16S/c1-2-3-4-5-6-7-8/h8H,2-7H2,1H3

InChI key

VPIAKHNXCOTPAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alif Chebbi et al.
FEMS microbiology letters, 362(14) (2015-06-19)
Hydrogen sulfide (H2S) and thiols (RSH) generated by the phosphate industry cause harmful effects on human health and quality of life. The present study aims to investigate and evaluate a bacterial strain CAT37 isolated from gas-washing wastewaters in terms of

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service