Skip to Content
Merck
All Photos(1)

Key Documents

H19859

Sigma-Aldrich

2-Hydroxybenzimidazole

97%

Synonym(s):

2(1H)-Benzimidazolone, 2-Hydroxy-benzimidazol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€35.80
25 G
€130.00

€35.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€35.80
25 G
€130.00

About This Item

Empirical Formula (Hill Notation):
C7H6N2O
CAS Number:
Molecular Weight:
134.14
Beilstein:
119866
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€35.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

>300 °C (lit.)

SMILES string

O=C1Nc2ccccc2N1

InChI

1S/C7H6N2O/c10-7-8-5-3-1-2-4-6(5)9-7/h1-4H,(H2,8,9,10)

InChI key

SILNNFMWIMZVEQ-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Bojanowska-Czajka et al.
Journal of radioanalytical and nuclear chemistry, 289(2), 303-314 (2011-01-01)
The radiolytic degradation of widely used fungicide, carbendazim, in synthetic aqueous solutions and industrial wastewater was investigated employing γ-irradiation. The effect of the absorbed dose, initial concentration and pH of irradiated solution on the effectiveness of carbendazim decomposition were investigated.
Xinjian Zhang et al.
PloS one, 8(10), e74810-e74810 (2013-10-08)
Carbendazim (methyl 1H-benzimidazol-2-yl carbamate) is one of the most widely used fungicides in agriculture worldwide, but has been reported to have adverse effects on animal health and ecosystem function. A highly efficient carbendazim-degrading bacterium (strain dj1-11) was isolated from carbendazim-contaminated
Vangelis Agouridas et al.
Chemical reviews, 119(12), 7328-7443 (2019-05-06)
The native chemical ligation reaction (NCL) involves reacting a C-terminal peptide thioester with an N-terminal cysteinyl peptide to produce a native peptide bond between the two fragments. This reaction has considerably extended the size of polypeptides and proteins that can
Miki Matsui et al.
Molecular pharmacology, 95(3), 294-302 (2019-01-10)
The hyperpolarization induced by intermediate-conductance Ca2+-activated K+ channel (KCa3.1) activation increases the driving force for Ca2+ influx, which generally promotes cell proliferation, migration, and cytokine production in immunocompetent cells. Interleukin-10 (IL-10) from tumor-infiltrating lymphocytes and macrophages, lymphoma, and carcinoma cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service