Skip to Content
Merck
All Photos(2)

Key Documents

GF96972216

Silver

foil, light tested, 150x150mm, thickness 0.05mm, annealed, 99.95+%

Synonym(s):

Silver, AG000305

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12141740
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

≥99.95%

form

foil

manufacturer/tradename

Goodfellow 969-722-16

resistivity

1.59 μΩ-cm, 20°C

bp

2212 °C (lit.)

mp

960 °C (lit.)

density

10.49 g/cm3 (lit.)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiping Zhuang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 72(5), 954-958 (2009-02-03)
Surface-enhanced Raman scattering (SERS) of 4,4'-azopyridine (AZPY) on silver foil substrate was measured under 1064nm excitation lines. Density-functional theory (DFT) methods were used to calculate the structure and vibrational spectra of models such as Ag-AZPY, Ag(4)-AZPY and Ag(6)-AZPY complexes with
A Rosenblatt et al.
Journal of dental research, 88(2), 116-125 (2009-03-13)
The antimicrobial use of silver compounds pivots on the 100-year-old application of silver nitrate, silver foil, and silver sutures for the prevention and treatment of ocular, surgical, and dental infections. Ag(+) kills pathogenic organisms at concentrations of <50 ppm, and
C Gonzalez et al.
The Annals of otology, rhinology, and laryngology, 99(7 Pt 1), 553-555 (1990-07-01)
The erbium:yttrium aluminum garnet (Er:YAG) laser operates in the infrared spectrum at a wavelength of 2.94 microns. The absorption coefficient for water at this wavelength is approximately 10 times that of the carbon dioxide laser. It is anticipated that this
Mukul Das et al.
Food additives and contaminants, 22(12), 1219-1223 (2005-12-17)
The use of silver foils in various food preparations is a common practice in Middle Eastern and South East Asian countries. The FAO/WHO Joint Expert Committee on Food Additives (JECFA) has included silver in the list of food additives, but
Cheng Yang et al.
Physical chemistry chemical physics : PCCP, 12(43), 14459-14461 (2010-10-01)
This communication describes a new surface-enhanced Raman scattering (SERS) active silver substrate prepared by iodination of the evaporated silver foil. After iodination, the morphology of the silver substrate undergoes a self-evolution process in which it displays accordingly the UV-vis absorption

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service