Skip to Content
Merck
All Photos(1)

Key Documents

GF87410819

Lithium fluoride

lump, 10 mm max. lump size, weight 10 g, high purity 99.995%

Synonym(s):

LI516101

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€340.00

€340.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€340.00

About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

€340.00


Check Cart for Availability

Request a Bulk Order

Assay

99.995%

form

lumps

manufacturer/tradename

Goodfellow 874-108-19

weight

10 g

max. lump size

10 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service