Skip to Content
Merck
All Photos(1)

Documents

GF86777746

Alumina/Silica/Boria

fiber, tex number 680, Al2O3 62%/SiO2 24%/B2O3 14%, number of filaments, length 5 m

Synonym(s):

AL625725

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11181511
NACRES:
NA.23

form

fibers

manufacturer/tradename

Goodfellow 867-777-46

L

5 m

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service