GF85707513
Palladium
wire reel, 0.5m, diameter 1.0mm, hard, 99.95%
Synonym(s):
Palladium, PD005150
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Assay
≥99.95%
form
wire
manufacturer/tradename
Goodfellow 857-075-13
resistivity
9.96 μΩ-cm, 20°C
bp
2970 °C (lit.)
mp
1554 °C (lit.)
density
12.02 g/cm3 (lit.)
SMILES string
[Pd]
InChI
1S/Pd
InChI key
KDLHZDBZIXYQEI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chemical Society reviews, 41(10), 3929-3968 (2012-03-27)
A survey highlighting the most recent palladium catalytic systems produced and their performances for progress in direct synthesis of indole backbones by heterocarbocyclization of reactive substrates is provided. The discussion is developed in relation with the principles of sustainable chemistry
Chemical & pharmaceutical bulletin, 60(3), 285-299 (2012-03-03)
It is known that propargylic compounds having an ester and a halide at the propargylic positions react with palladium complexes leading to π-propargylpalladium and allenylpalladium complexes, which cause various transformations in the presence of the reactants. The aim of the
Molecules (Basel, Switzerland), 18(6), 6173-6192 (2013-05-28)
The palladium (II)-catalysed reactions of alkenols and aminoalkenols such as oxycarbonylations or bicyclisations are powerful methods for the construction of oxygen and nitrogen-containing heterocyclic compounds. This review highlights recent progress in the development of the asymmetric palladium(II)-catalysed Wacker-type cyclisations of
Synthesis of heterocycles via palladium-catalyzed carbonylations.
Chemical reviews, 113(1), 1-35 (2012-10-09)
Update 1 of: Synthesis and functionalization of indoles through palladium-catalyzed reactions.
Chemical reviews, 111(5), PR215-PR283 (2011-05-12)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service