Skip to Content
Merck
All Photos(2)

Key Documents

GF84638065

Titanium

powder, 150 max. part. size (micron), weight 1000 g, purity 99.5%

Synonym(s):

TI006020

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€1,080.00

€1,080.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€1,080.00

About This Item

Empirical Formula (Hill Notation):
Ti
CAS Number:
Molecular Weight:
47.87
MDL number:
UNSPSC Code:
12141746
PubChem Substance ID:
NACRES:
NA.23

€1,080.00


Check Cart for Availability

Request a Bulk Order

Assay

99.5%

form

powder

autoignition temp.

860 °F

manufacturer/tradename

Goodfellow 846-380-65

resistivity

42.0 μΩ-cm, 20°C

weight

1000 g

max. part. size

150 (micron)

bp

3287 °C (lit.)

mp

1660 °C (lit.)

density

4.5 g/mL at 25 °C (lit.)

SMILES string

[Ti]

InChI

1S/Ti

InChI key

RTAQQCXQSZGOHL-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hello, I'm looking for the powder density of this product, if it is wide or narrow? Also is this recommended/safe/compatible for Directed energy deposition with laser powder fed systems such as an Optomec LENS 500, which needs 45 to 150 microns.

    1 answer
    1. The determination of powder density is not a quality specification for the product and, thus, has not been determined. There is no data available showing the product is suitable for use in directed energy deposition with laser powder fed systems.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service