Skip to Content
Merck
All Photos(2)

Documents

GF82419408

Platinum

foil, 50x50mm, thickness 0.125mm, as rolled, 99.95%

Synonym(s):

Platinum, PT000251

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.95%

form

foil

manufacturer/tradename

Goodfellow 824-194-08

resistivity

10.6 μΩ-cm, 20°C

size × thickness

50 x 50 mm × 0.125 mm

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasar F Mansour et al.
Acta odontologica Scandinavica, 64(4), 231-236 (2006-07-11)
To quantify luting cement at the crown-tooth interface of esthetic crowns fabricated using four different techniques and two methods of excess cement removal. Four methods of crown fabrication were used: the feldspathic porcelain and platinum foil technique, the feldspathic porcelain
A Medici et al.
Steroids, 66(2), 63-69 (2001-01-09)
Regioselectivity in the anodic electrochemical oxidation of cholic acid with different anodes is described. The oxidation with PbO(2) anode affords the dehydrocholic acid in quantitative yield after 22 h. 3alpha,12alpha-Dihydroxy-7-oxo-5beta-cholan-24-oic acid (59%) and 3alpha-hydroxy-7,12-dioxo-5beta-cholan-24-oic acid (51%) are obtained stopping the
Christian Henle et al.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference, 2008, 4208-4211 (2009-01-24)
A laser technology for manufacturing implantable electrode arrays, using spin-on polydimethylsiloxane (PDMS) and platinum foil as materials, was investigated in respect to its scaling limitations. The following aspects were analyzed: Minimal width and centre-to-centre distance of platinum tracks, the ability
Minimally invasive makeover: platinum foil technique.
Steve Ballis et al.
Dentistry today, 29(2), 146-146 (2010-03-04)
Using platinum foil techniques to create natural tooth color.
Luke Kahng
Practical procedures & aesthetic dentistry : PPAD, 20(8), 479-481 (2008-11-11)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service