Skip to Content
Merck
All Photos(2)

Documents

GF76383254

Indium

wire reel, 2m, diameter 0.5mm, as drawn, 99.999%

Synonym(s):

Indium, IN005120

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
In
CAS Number:
Molecular Weight:
114.82
MDL number:
UNSPSC Code:
12141719
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 25 °C)

Assay

99.999%

form

wire

manufacturer/tradename

Goodfellow 763-832-54

resistivity

8.37 μΩ-cm

L × diam.

2 m × 0.5 mm

mp

156.6 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

[In]

InChI

1S/In

InChI key

APFVFJFRJDLVQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methodologic and basic aspects of indium-111 platelets.
J H Joist et al.
Seminars in thrombosis and hemostasis, 9(2), 86-99 (1983-04-01)
New generation of palladium-indium-silver dental cast alloys: a review.
M L Malhotra
Trends & techniques in the contemporary dental laboratory, 9(4), 65-68 (1992-05-01)
Uwe Schneider et al.
Accounts of chemical research, 45(8), 1331-1344 (2012-05-26)
The development of innovative metal catalysis for selective bond formation is an important task in organic chemistry. The group 13 metal indium is appealing for catalysis because indium-based reagents are minimally toxic, selective, and tolerant toward various functional groups. Among
Alex N Eberle et al.
Journal of molecular recognition : JMR, 16(5), 248-254 (2003-10-03)
Following the first synthesis of tritiated alpha-melanocyte-stimulating hormone (alpha-MSH, alpha-melanotropin) in 1974 by Medzihradszky et al., several alpha-MSH analogs were designed containing between 2 and 12 tritium atoms, the latter of which displayed a specific radioactivity of 12.21 GBq/micromol (330
Akiyo Tanaka
Toxicology and applied pharmacology, 198(3), 405-411 (2004-07-28)
Gallium arsenide (GaAs), indium arsenide (InAs), and aluminium gallium arsenide (AlGaAs) are semiconductor applications. Although the increased use of these materials has raised concerns about occupational exposure to them, there is little information regarding the adverse health effects to workers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service