Skip to Content
Merck
All Photos(2)

Key Documents

GF75380430

Zinc

foil, 300x300mm, thickness 2.0mm, as rolled, 99.95+%

Synonym(s):

Zinc, ZN000370

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 487 °C)

Assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 753-804-30

resistivity

5.8 μΩ-cm, 20°C

size × thickness

300 x 300 mm × 2.0 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service