Skip to Content
Merck
All Photos(2)

Key Documents

GF75296469

Cadmium

foil, 25mm disks, thickness 0.125mm, as rolled, 99.99+%

Synonym(s):

Cadmium, CD000230

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cd
CAS Number:
Molecular Weight:
112.41
Beilstein:
8137359
MDL number:
UNSPSC Code:
12141707
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.99%

form

foil

manufacturer/tradename

Goodfellow 752-964-69

resistivity

7.27 μΩ-cm, 22°C

diam. × thickness

25 m × 0.125 mm

bp

765 °C (lit.)

mp

320.9 °C (lit.)

density

8.65 g/mL at 25 °C (lit.)

SMILES string

[Cd]

InChI

1S/Cd

InChI key

BDOSMKKIYDKNTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Nakamura et al.
Radioisotopes, 28(11), 675-680 (1979-11-01)
Formation cross sections and thick-target yields were studied in detail for alpha reactions and helium-3 reactions of natural cadmium in the energy range 10-40 MeV in order to determine the optimum condition for the production of medical use 11In. The
Uroš Cvelbar et al.
Nanotechnology, 19(40), 405605-405605 (2008-10-08)
Plasma-assisted synthesis of nanostructures is one of the most precise and effective approaches used in nanodevice fabrication. Here we report on the innovative approach of synthesizing nanostructured cadmium oxide films on Cd substrates using a reactive oxygen plasma-based process. Under
Jungsoo Kim et al.
Environmental monitoring and assessment, 186(4), 2321-2327 (2014-01-11)
In birds, concentrations of heavy metal contaminants were associated with their diet and environmental quality. This study presents concentrations of lead and cadmium in feathers and the relationship between concentrations in the feather and stomach content of three species of
S E Dal-Medico et al.
Ecotoxicology and environmental safety, 104, 168-174 (2014-04-01)
A drastic amphibian decline has been observed worldwide, which can be attributed (among other factors) to exposure to pollutants. Considering that cadmium corresponds to the most rapidly increasing trace metal in the environment, the aim of this work was to
Burcu Yeşilbudak et al.
Bulletin of environmental contamination and toxicology, 92(5), 546-550 (2014-02-15)
Accumulation of cadmium in gill, liver, muscle and kidney tissues of Cyprinus carpio and Oreochromis niloticus were investigated in fish exposed to 0.5 ppm cadmium over 1, 15 and 30 days under controlled laboratory conditions. Tissue accumulation of the metal was measured

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service