Skip to Content
Merck
All Photos(1)

Key Documents

GF70011021

Polyethylene - High Density

(HDPE), granule, 2-4 mm nominal granule size, weight 0.5 kg

Synonym(s):

ET326310

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€215.00

€215.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
€215.00

About This Item

UNSPSC Code:
12352100
NACRES:
NA.23

€215.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

granular

manufacturer/tradename

Goodfellow 700-110-21

weight

0.5 kg

nominal granule size

2-4 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. what is the molecular weight of the HDPE?

    1 answer
    1. The molecular weight for this product is not determined.

      Helpful?

  2. What color are the granules?

    1 answer
    1. This material is listed as 'natural' in color. The typical color of HDPE ranges from translucent to opaque white.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service