GF62139876
Boron carbide-hot-pressed
sheet, thickness 4.0 mm, size 50 × 50 mm
Select a Size
About This Item
Recommended Products
form
sheet
manufacturer/tradename
Goodfellow 621-398-76
size
50 × 50 mm
thickness
4.0 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service