Skip to Content
Merck
All Photos(2)

Key Documents

GF57344654

Tin

foil, 6mm disks, thickness 0.5mm, as rolled, 99.99+%

Synonym(s):

Tin, SN000420

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sn
CAS Number:
Molecular Weight:
118.71
MDL number:
UNSPSC Code:
12141745
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.99%

form

foil

manufacturer/tradename

Goodfellow 573-446-54

resistivity

11 μΩ-cm, 20°C

diam. × thickness

6 mm × 0.5 mm

bp

2270 °C (lit.)

mp

231.9 °C (lit.)

density

7.310 g/mL at 25 °C (lit.)

SMILES string

[Sn]

InChI

1S/Sn

InChI key

ATJFFYVFTNAWJD-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianan Liu et al.
Animal reproduction science, 134(3-4), 197-202 (2012-08-28)
Cryopreservation of ovarian tissue has been the only effective way of ex situ conservation of female germplasm in avian species. A novel needle-in-straw (NIS) vitrification method was developed to store tissue in straws instead of cryovials. Fragments of ovarian tissue
J Strang et al.
Addiction (Abingdon, England), 92(6), 673-683 (1997-06-01)
The history of heroin smoking and the subsequent development and spread of 'chasing the dragon' are examined. The first heroin smoking originated in Shanghai in the 1920s and involved use of porcelain bowls and bamboo tubes, thereafter spreading across much
Andrey A Gurtovenko et al.
The Journal of chemical physics, 130(21), 215107-215107 (2009-06-11)
The electrostatic properties of lipid membranes are of profound importance as they are directly associated with membrane potential and, consequently, with numerous membrane-mediated biological phenomena. Here we address a number of methodological issues related to the computation of the electrostatic
Mercury contamination removed with tin foil.
R Grajower et al.
Operative dentistry, 9(3), 101-104 (1984-01-01)
K A Winship
Adverse drug reactions and acute poisoning reviews, 7(1), 19-38 (1988-01-01)
Inorganic tin salts are poorly absorbed and rapidly excreted in the faeces; as a result they have a low toxicity. Only about 5 per cent is absorbed from the gastrointestinal tract, widely distributed in the body, then excreted by the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service