Skip to Content
Merck
All Photos(2)

Key Documents

GF54906385

Cobalt

wire reel, 0.025m, diameter 2.0mm, hard, 99.99+%

Synonym(s):

Cobalt, CO005130

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€437.00

€437.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
€437.00

About This Item

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
MDL number:
UNSPSC Code:
12141710
PubChem Substance ID:
NACRES:
NA.23

€437.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Assay

99.99%

form

wire

manufacturer/tradename

Goodfellow 549-063-85

resistivity

6.24 μΩ-cm, 20°C

L × diam.

0.025 m × 2.0 mm

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dennis J Paustenbach et al.
Clinical toxicology (Philadelphia, Pa.), 52(2), 98-112 (2013-11-22)
INTRODUCTION. There has been some recent concern regarding possible systemic health effects resulting from elevated blood cobalt concentrations in patients with cobalt containing hip implants. To date there are no blood cobalt criteria to help guide physicians when evaluating an
Kenneth M Unice et al.
Chemico-biological interactions, 216, 53-74 (2014-04-15)
An updated biokinetic model for human exposures to cobalt (Co) was developed based on a comprehensive set of human pharmacokinetics data collected from five male and five female volunteers who ingested ∼1 mg Co/day of a Co supplement for 3
Hao-Di Wu et al.
Journal of nanoscience and nanotechnology, 14(6), 4097-4100 (2014-04-18)
Cocrystal nanofibres of cobalt octaethylporphyrin and tetracyanoquinodimethane were prepared by a facile solution method and fully characterized by SEM, AFM, XRD, Raman, EDX, and UV-vis-NIR. The as-prepared cocrystal nanofibres had smooth surfaces and uniform dimension. When incorporated into prototype devices
Kazuhiro Yamada
Metal ions in life sciences, 13, 295-320 (2014-01-29)
The primarily function of cobalt in humans is based on its role in cobalamin (Cbl, vitamin B12). Therefore, this chapter will focus on the physiological roles of Cbl and the importance of cobalt in human health. Cbl acts as the
K Praveena et al.
Journal of nanoscience and nanotechnology, 14(6), 4371-4376 (2014-04-18)
The Cobalt ferrite (CoFe2O4) powders were synthesized by Co-precipitation method. The as prepared ferrite powders were incorporated into a polyaniline matrix at various volumetric ratios. The as prepared composites of ferrite and polyaniline powders were characterized using X-ray diffraction (XRD)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service