Skip to Content
Merck
All Photos(1)

Key Documents

GF48489430

Tellurium

lump, 25 mm max. lump size, weight 100 g, high purity 99.999%

Synonym(s):

TE006100

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€663.00

€663.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€663.00

About This Item

UNSPSC Code:
12141913
NACRES:
NA.23

€663.00


Check Cart for Availability

Request a Bulk Order

Assay

99.999%

form

lumps

manufacturer/tradename

Goodfellow 484-894-30

weight

100 g

max. lump size

25 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 4 - Lact. - Repr. 1B - Skin Sens. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service