GF47218530
Alumina/Silica/Boria
fiber, Al2O3 62%/SiO2 24%/B2O3 14%, tex number 680, length 10 m, number of filaments
Synonym(s):
AL625725
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
fibers
manufacturer/tradename
Goodfellow 472-185-30
L
10 m
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service