Skip to Content
Merck
All Photos(1)

Key Documents

GF38890165

Superconducting composite wire alloy, NbTi+Cu

0.5m wire, 0.4mm diameter, copper/superconductor ratio 3:1 by volume

Synonym(s):

NB015100

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€221.00

€221.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
€221.00

About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

€221.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

wire

manufacturer/tradename

Goodfellow 388-901-65

L × diam.

0.5 m × 0.4 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the critical temperature? Thanks for respond

    1 answer
    1. The critical temperature for this product is not recorded. However, the website states the manufacturer as Goodfellow in the "Properties" section of the product page. Goodfellow's product number to this item is 388-901-65. You may find it helpful to view the pdf Goodfellow provides that lists the physical characteristics and thermal properties of all the chemicals used in their products. This product contains Niobium, Titanium, and Copper. Pages 192, 195, and 198 list those chemicals and their properties.

      Please see the manufacture Content pdf: https://www.goodfellow.co.kr/pub/media/pdf/Goodfellow-Product-List-Final.pdf

      There is a specific value assigned to each superconductor metal - for Nb it is 9.25 Kelvin and for Ti it is 0.40 Kelvin. The critical temperature for a NbTi Alloy superconductor is approximately 10 Kelvin. More information on the development of this superconductor material can be found in this reference: https://www.sciencedirect.com/science/article/abs/pii/0011227587900579?via%3Dihub

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service