Skip to Content
Merck
All Photos(2)

Key Documents

GF36412871

Lead

wire reel, 5m, diameter 2.0mm, as drawn, 99.99+%

Synonym(s):

Lead, PB005140

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€1,460.00

€1,460.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
€1,460.00

About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
MDL number:
UNSPSC Code:
12141723
PubChem Substance ID:
NACRES:
NA.23

€1,460.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Assay

99.99%

form

wire

manufacturer/tradename

Goodfellow 364-128-71

resistivity

20.65 μΩ-cm

L × diam.

5 m × 2.0 mm

bp

1740 °C (lit.)

mp

327.4 °C (lit.)

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reply: Cognitive declines after deep brain stimulation are likely to be attributable to more than caudate penetration and lead location.
Karsten Witt et al.
Brain : a journal of neurology, 137(Pt 5), e275-e275 (2014-02-20)
Qinglin Chen et al.
Ecotoxicology and environmental safety, 102, 93-99 (2014-03-04)
The experiment was conducted to investigate the effects of single and combined pollution from different concentrations of atrazine (field rate, FR, 2.0 mg kg(-1) and 5 times FR, 10 mg kg(-1)) and lead (Pb) (300 mg kg(-1) and 600 mg
Chamani P M Marasinghe Wadige et al.
Aquatic toxicology (Amsterdam, Netherlands), 149, 83-93 (2014-03-01)
Lead entering aquatic ecosystems adsorbs to sediments and has the potential to cause adverse effects on the health of benthic organisms. To evaluate the freshwater bivalve Hyridella australis as a bioindicator for sediment toxicity, their exposure-dose and response to lead
Paraskevi Malea et al.
Ecotoxicology and environmental safety, 104, 175-181 (2014-04-01)
The effects of lead uptake on microtubule integrity and cell viability in intermediate-juvenile leaf blades of the seagrass Cymodocea nodosa were investigated under laboratory conditions in increasing exposure concentrations (0.1, 0.25, 0.5, 5, 10, 20 and 40mg/L). Uptake kinetics was
B Ali et al.
Ecotoxicology and environmental safety, 102, 25-33 (2014-03-04)
The role of hydrogen sulfide (H2S) in alleviating lead (Pb) induced stress in oilseed rape (Brassica napus L.) was studied under laboratory conditions. Plants were grown hydroponically in greenhouse conditions under three levels (0, 100, and 400 µM) of Pb

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service