Skip to Content
Merck
All Photos(2)

Key Documents

GF31925213

Carbon

tube, outside diameter 3.18 mm, length 150 mm, wall thickness 0.8 mm

Synonym(s):

Activated charcoal, C 007201, Charcoal activated

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€492.00

€492.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€492.00

About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
12141908
PubChem Substance ID:
NACRES:
NA.23

€492.00


Check Cart for Availability

Request a Bulk Order

vapor pressure

<0.1 mmHg ( 20 °C)

form

tubes

autoignition temp.

842 °F

manufacturer/tradename

Goodfellow 319-252-13

resistivity

1375 μΩ-cm, 20°C (graphite)

L

150 mm

O.D.

3.18 mm

wall thickness

0.8 mm

mp

3550 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mengmei Zhang et al.
Journal of hazardous materials, 388, 122064-122064 (2020-01-19)
Chromium is commonly found in the flue gases and ashes of Municipal Solid Waste Incineration. It has been reported as an active catalyst for the formation of polychlorinated dibenzo-p-dioxins and dibenzofurans (PCDD/F) during de novo tests, yet its specific mode
Jaclyn Gowen Kalmar et al.
Analytical and bioanalytical chemistry, 412(27), 7569-7579 (2020-08-28)
The analysis of N-linked glycans using liquid chromatography and mass spectrometry (LC-MS) presents significant challenges, particularly owing to their hydrophilic nature. To address these difficulties, a variety of derivatization methods have been developed to facilitate improved ionization and detection sensitivity.
Devika Sasikumar et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(9), 2060-2066 (2019-12-15)
The conversion of solar energy to thermal, chemical, or electrical energy attracts great attention in chemistry and physics. There has been a considerable effort for the efficient extraction of photons throughout the entire solar spectrum. In this work light energy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service