GF31674955
Gold-iron alloy, Au99.93Fe0.07
1m insulated wire, 0.1mm conductor diameter, 0.01mm insulation thickness, ptfe (polytetrafluoroethylene) insulation
Synonym(s):
AU075840
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
description
ptfe (polytetrafluoroethylene) insulation
form
wire (insulated)
manufacturer/tradename
Goodfellow 316-749-55
L
1 m
conductor diam.
0.1 mm
insulation thickness
0.01 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service