Skip to Content
Merck
All Photos(1)

Key Documents

GF30719471

Stainless steel - AISI 316l

powder, Fe/Cr18%/Ni 10%/Mo 3%, mean particle size 3 micron, weight 20 g, condition atomized

Synonym(s):

FF216030

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€328.00

€328.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€328.00

About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

€328.00


Check Cart for Availability

Request a Bulk Order

form

powder

manufacturer/tradename

Goodfellow 307-194-71

weight

20 g

mean particle size

3 μm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Flam. Sol. 1 - Self-heat. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service