Skip to Content
Merck
All Photos(1)

Documents

GF27696213

Nickel silver (18) alloy, Cu65Ni18Zn17

500mm tube, 6.35mm outside diameter, 0.5mm wall thickness, 5.35mm inside diameter, hard

Synonym(s):

CZ057750

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 276-962-13

L × wall thickness

500 mm × 0.5 mm

O.D. × I.D.

6.35 mm × 5.35 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service