GF27692747
Polyethylene - Low Density
(LDPE), rod, diam. 101.6 mm, L 500 mm, natural
Synonym(s):
ET317990
Select a Size
€1,080.00
Select a Size
About This Item
€1,080.00
form
rod
manufacturer/tradename
Goodfellow 276-927-47
L
500 mm
diam.
101.6 mm
color
natural
Related Categories
General description
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service