GF20441214
Inconel® alloy 600 - heat resisting alloy, Ni72Cr16Fe8
foil, 5m coil, 76mm coil width, 0.1mm thickness, annealed
Synonym(s):
NI020206
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
annealed
foil (coil)
manufacturer/tradename
Goodfellow 204-412-14
L × W × thickness
5 m × 76 mm × 0.1 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
INCONEL is a registered trademark of Special Metals Corporation
Inconel is a registered trademark of Special Metals Corporation
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service