Skip to Content
Merck
All Photos(2)

Documents

GF17713537

Lead

foil, 500x500mm, thickness 3.0mm, as rolled, 99.95%

Synonym(s):

Lead, PB000472

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
MDL number:
UNSPSC Code:
12141723
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 177-135-37

resistivity

20.65 μΩ-cm

size × thickness

300 x 300 mm × 3.0 mm

bp

1740 °C (lit.)

mp

327.4 °C (lit.)

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Bourgeois et al.
Health physics, 62(6), 546-552 (1992-06-01)
A significant amount of radiation escapes the rear of an image receptor in conventional dental radiographic procedures despite the placement of a lead-containing backing at the rear of the film pack. The purpose of this investigation was to place additional
Motoaki Kato et al.
Nihon Hoshasen Gijutsu Gakkai zasshi, 61(8), 1158-1169 (2005-09-01)
In projection radiography, stationary grids are indispensable accessories to the improvement of diagnostic imaging. On the other hand, they are becoming one of the issues facing digital image processing. The lead foil that composes the grid can produce moiré on
D W Rogers
Medical physics, 26(4), 533-537 (1999-05-05)
Data are presented to allow the photon beam quality specifier being used in the new AAPM TG-51 protocol, %dd(10)x, to be extracted from depth-dose data measured with a 1 mm lead foil either 50 cm or 30 cm from the
X A Li et al.
Medical physics, 26(7), 1245-1251 (1999-08-06)
The purpose of this work is to study the dose enhancement by a thin foil (thickness of 0.2-4 mm) of high-Z material in a water phantom, irradiated by high-energy photon beams. EGS4 Monte Carlo technique was used. Perturbations on the
I Sewerin
Tandlaegebladet, 94(12), 483-486 (1990-09-01)
The lead foil of dental x-ray film packets may cause a number of artifacts; especially well known are the "tractor treads" and "herringbone patterns" following a backward filmpositioning in the patient's mouth. In the present paper a specific and seldomly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service