Skip to Content
Merck
All Photos(2)

Key Documents

GF14472514

Tungsten

foil, 0.2m coil, thickness 0.1mm, coil width .6mm, as rolled, 99.95%

Synonym(s):

Tungsten, W 000278, W

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€186.00

€186.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
€186.00

About This Item

Empirical Formula (Hill Notation):
W
CAS Number:
Molecular Weight:
183.84
MDL number:
UNSPSC Code:
12141747
PubChem Substance ID:
NACRES:
NA.23

€186.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Assay

99.50%

form

foil

manufacturer/tradename

Goodfellow 144-725-14

resistivity

4.9 μΩ-cm, 20°C

L × W × thickness

0.2 m × 0.6 mm × 0.1 mm

bp

5660 °C (lit.)

mp

3410 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[W]

InChI

1S/W

InChI key

WFKWXMTUELFFGS-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Congshang Wan et al.
Ultramicroscopy, 119, 106-110 (2011-11-15)
Thermionic electron emission from 200 to 500 nm thick coatings of scandium oxide on tungsten foil have been examined in thermionic emission microscopy, spectroscopic photoelectron microcopy, synchrotron radiation and ultraviolet photoelectron spectroscopy (UPS). A clear dependence of the scandium oxide-W
J R Marbach et al.
Physics in medicine and biology, 26(3), 435-443 (1981-05-01)
Curves relating beam energy, scattering foil thickness, central-axis depth-dose, and beam flatness have been generated using data taken on a Siemens 200A betatron. The curve set allows a single combination of tungsten foil thickness and electron beam energy to be
Dae Yup Han et al.
International journal of radiation oncology, biology, physics, 89(3), 666-673 (2014-04-23)
To demonstrate that utilization of the direction-modulated brachytherapy (DMBT) concept can significantly improve treatment plan quality in the setting of high-dose-rate (HDR) brachytherapy for cervical cancer. The new, MRI-compatible, tandem design has 6 peripheral holes of 1.3-mm diameter, grooved along
Gijsbert B van der Voet et al.
Military medicine, 172(9), 1002-1005 (2007-10-17)
Tungsten and tungsten compounds are considered toxicologically relatively safe. Concern regarding the potential health and environmental effects of depleted uranium and lead in military applications has lead many countries to explore the possibility of applying toxicologically safer metals. Heavy metal
Carlos D Brondino et al.
Current opinion in chemical biology, 10(2), 109-114 (2006-02-17)
Mononuclear molybdenum and tungsten are found in the active site of a diverse group of enzymes that, in general, catalyze oxygen atom transfer reactions. Enzymes of the xanthine oxidase family are the best-characterized mononuclear Mo-containing enzymes. Several 3D structures of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service