Skip to Content
Merck
All Photos(1)

Documents

GF13723714

Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3

50m insulated wire, 0.075mm conductor diameter, 0.018mm insulation thickness, ptfe (polytetrafluoroethylene) insulation

Synonym(s):

FE245810

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

description

ptfe (polytetrafluoroethylene) insulation

form

wire (insulated)

manufacturer/tradename

Goodfellow 137-237-14

L

50 m

conductor diam.

0.075 mm

insulation thickness

0.018 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service