Skip to Content
Merck
All Photos(2)

Key Documents

GF11817907

Nickel

powder, max. particle size 150 micron, weight 200 g, high purity 99.99+%

Synonym(s):

NI006035

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€2,010.00

€2,010.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€2,010.00

About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
MDL number:
UNSPSC Code:
12141729
PubChem Substance ID:
NACRES:
NA.23

€2,010.00


Check Cart for Availability

Request a Bulk Order

Assay

≥99.99%

form

powder

manufacturer/tradename

Goodfellow 118-179-07

resistivity

6.97 μΩ-cm, 20°C

weight

200 g

max. part. size

150 μm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract,Skin

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service