Skip to Content
Merck
All Photos(2)

Key Documents

GF10931947

Silver

foil, 0.5m coil, thickness 0.01mm, 99.95+%

Synonym(s):

Silver, AG000220

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€2,340.00

€2,340.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
€2,340.00

About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12141740
PubChem Substance ID:
NACRES:
NA.23

€2,340.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 109-319-47

resistivity

1.59 μΩ-cm, 20°C

L × thickness

0.5 m × 0.01 mm

bp

2212 °C (lit.)

mp

960 °C (lit.)

density

10.49 g/cm3 (lit.)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fan Yang et al.
Talanta, 84(4), 1099-1106 (2011-05-03)
In this paper, a compact and inexpensive light emitting diode induced fluorescence (LED-IF) detector with simplified optical configuration was developed and assembled in an integrated microfluidic device for microscale electrophoresis. The facile detector mainly consisted of an LED, a focusing
R D Stickrod et al.
Rapid communications in mass spectrometry : RCM, 14(14), 1266-1268 (2000-08-05)
Nitrate-delta(15)N from groundwater samples is determined on an inorganic nitrate derivative using automated, continuous-flow elemental analyzer/isotope ratio mass spectrometry (EA/IRMS). Nitrate is extracted and concentrated based on a recently published ion-exchange resin method. Freeze-dried AgNO(3) (0.5-1.5 mg) is packed in
Ken-ichiro Matsumoto et al.
Biological & pharmaceutical bulletin, 28(11), 2029-2034 (2005-11-08)
A novel screening experiment, to find radioactive probes for non-invasive measurements of physiological functions in experimental animals, was tested using the in vivo multitracer analysis technique. The details of the efficiency of the detector settings used in the in vivo
Zhiping Zhuang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 67(2), 509-516 (2006-09-22)
The molecular geometry and vibrational frequencies of 4,4'-bipyridine (BPE) in the ground state were calculated using density functional theory (DFT) methods (B3LYP) with 6-31++G(d,p) basis set. The optimized geometric bond lengths and bond angles are obtained by DFT employing the
IX. Note on Silver Foil in Surgery.
J S Lewis
Annals of surgery, 50(4), 793-796 (1909-10-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service