G4780
D(+)-Glyceraldehyde solution
≥70 wt. % in H2O
Synonym(s):
(R)-(+)-Glyceraldehyde, D-Glyceraldehyde solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HOCH2CH(OH)CHO
CAS Number:
Molecular Weight:
90.08
Beilstein:
1720474
EC Number:
MDL number:
UNSPSC Code:
12352200
optical activity
[α]23/D +9.4°, c = 2 in H2O
concentration
≥70 wt. % in H2O
refractive index
n20/D 1.494
SMILES string
OC[C@@H](O)C=O
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service