Skip to Content
Merck
All Photos(1)

Key Documents

F7407

Sigma-Aldrich

4-Fluorobenzyl alcohol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€36.10

€36.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€36.10

About This Item

Linear Formula:
FC6H4CH2OH
CAS Number:
Molecular Weight:
126.13
Beilstein:
1446930
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€36.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.507 (lit.)

density

1.156 g/mL at 25 °C (lit.)

SMILES string

OCc1ccc(F)cc1

InChI

1S/C7H7FO/c8-7-3-1-6(5-9)2-4-7/h1-4,9H,5H2

InChI key

GEZMEIHVFSWOCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rianne E van Outersterp et al.
The Analyst, 145(18), 6162-6170 (2020-09-15)
Distinguishing positional isomers, such as compounds having different substitution patterns on an aromatic ring, presents a significant challenge for mass spectrometric analyses and is a frequently encountered difficulty in, for example, drug metabolism research. In contrast to mass spectrometry, IR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service