Skip to Content
Merck
All Photos(1)

Documents

F4061

Sigma-Aldrich

Fmoc-Lys(Mca)-OH

Synonym(s):

N-α-Fmoc-N-ε-7-methoxycoumarin-4-acetyl-L-Lys-OH, N-α-Fmoc-N-ε-7-methoxycoumarin-4-acetyl-L-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H32N2O8
CAS Number:
Molecular Weight:
584.62
UNSPSC Code:
12352200
NACRES:
NA.26

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

−20°C

InChI

1S/C33H32N2O8/c1-41-21-13-14-22-20(17-31(37)43-29(22)18-21)16-30(36)34-15-7-6-12-28(32(38)39)35-33(40)42-19-27-25-10-4-2-8-23(25)24-9-3-5-11-26(24)27/h2-5,8-11,13-14,17-18,27-28H,6-7,12,15-16,19H2,1H3,(H,34,36)(H,35,40)(H,38,39)/t28-/m0/s1

InChI key

BWJJONDPLYEAMC-NDEPHWFRSA-N

Application

Fmoc-Lys(Mca)-OH can be used as a building block to prepare fluorescent labeled peptides by solid-phase peptide synthesis. It is also used to synthesize luminescent lanthanide(III)-based chelates for spectroscopic applications, in particular, to label folded proteins or DNA.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

On-resin assembly of a linkerless lanthanide (III)-based luminescence label and its application to the total synthesis of site-specifically labeled mechanosensitive channels
Becker C FW, et al.
Bioconjugate Chemistry, 15(5), 1118-1124 (2004)
Alan R Katritzky et al.
Organic & biomolecular chemistry, 6(24), 4582-4586 (2008-11-29)
N(alpha)-Fmoc-N(epsilon)-[(7-methoxycoumarin-4-yl)acetyl]-L-lysine (N(alpha)-Fmoc-L-Lys(Mca)-OH) 3 is conveniently prepared by benzotriazole methodology (52% over two steps). N-Acylbenzotriazoles Mca-Bt 2, N(alpha)-Fmoc-L-Lys(Mca)-Bt 4, coumarin-3-ylcarbonyl (Cc)-Bt 5, N(alpha)-Fmoc-L-Lys(Cc)-Bt 7 and N(alpha)-(Cc)-L-Lys(Fmoc)-Bt 9 enable the efficient microwave enhanced solid-phase fluorescent labeling of peptides.
Christian F W Becker et al.
Bioconjugate chemistry, 15(5), 1118-1124 (2004-09-16)
A synthesis strategy for the on-resin assembly of luminescent lanthanide chelates from commercially available compounds was developed. Advantages of the approach include the absence of spacers between the metal ion and the attachment site, and the compatibility with typical chemical
Fluorescent labeling of peptides on solid phase
Katritzky AR, et al.
Organic & Biomolecular Chemistry, 6(24), 4582-4586 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service