Skip to Content
Merck
All Photos(1)

Documents

E45260

Sigma-Aldrich

Woodward′s reagent K

95%

Synonym(s):

2-Ethyl-5-phenylisoxazolium-3′-sulfonate, NEPIS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO4S
CAS Number:
Molecular Weight:
253.27
Beilstein:
4149224
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

reaction suitability

reaction type: Coupling Reactions

mp

220 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

CC[n+]1ccc(o1)-c2cccc(c2)S([O-])(=O)=O

InChI

1S/C11H11NO4S/c1-2-12-7-6-11(16-12)9-4-3-5-10(8-9)17(13,14)15/h3-8H,2H2,1H3

InChI key

MWOOKDULMBMMPN-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Bustos et al.
Journal of protein chemistry, 15(5), 467-472 (1996-07-01)
The reaction of Woordward's reagent K (WRK) with model amino acids and proteins has been analyzed. Our results indicate that WRK forms 340-nm-absorbing adducts with sulfhydryl- and imidazol-containing compounds, but not with carboxylic acid derivatives, in agreement with Liamas et
P Paoli et al.
The Biochemical journal, 328 ( Pt 3), 855-861 (1998-02-07)
The organ common-type (CT) isoenzyme of acylphosphatase is inactivated by Woodward's reagent K (WRK) (N-ethyl-5-phenylisoxazolium-3'-sulphonate) at pH6.0. The inactivation reaction follows apparent pseudo first-order kinetics. The dependence of the reciprocal of the pseudo first-order kinetic constant (kobs) on the reciprocal
Nelson Carvajal et al.
The protein journal, 23(3), 179-183 (2004-06-18)
Human liver arginase (EC 3.5.3.1) was totally inactivated by incubation with Woodward's reagent K (WRK). The inactivation followed pseudo-first-order kinetics, and the order of the inactivation was close to 1, consistent with reaction of one molecule of WRK with one
Hassan Faridnouri et al.
Bioelectrochemistry (Amsterdam, Netherlands), 82(1), 1-9 (2011-07-01)
This work describes the reaction mechanism for chemical modification of tyrosinase by Woodward's Reagent K and its covalent attachment to a glassy carbon electrode. The spectrophotometric studies revealed that the modification does not cause a significant structural change to tyrosinase.
Jongchan Woo et al.
Protein science : a publication of the Protein Society, 17(4), 725-735 (2008-03-25)
Renilla luciferase (RLUC) is a versatile tool for gene expression assays and in vivo biosensor applications, but its catalytic mechanism remains to be elucidated. RLUC is evolutionarily related to the alpha/beta hydrolase family. Its closest known homologs are bacterial dehalogenases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service