Skip to Content
Merck
All Photos(1)

Documents

E33408

Sigma-Aldrich

Ethyl isodehydracetate

99%

Synonym(s):

Ethyl 4,6-dimethyl-2-oxo-2H-pyran-5-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O4
CAS Number:
Molecular Weight:
196.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.515 (lit.)

bp

290-295 °C (lit.)

mp

17-18 °C (lit.)

density

1.167 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C1=C(C)OC(=O)C=C1C

InChI

1S/C10H12O4/c1-4-13-10(12)9-6(2)5-8(11)14-7(9)3/h5H,4H2,1-3H3

InChI key

FBPWNVQUVXSXKS-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Maione et al.
Steroids, 54(3), 313-320 (1989-09-01)
Condensation of C22 and C21 steroidal aldehydes with ethyl 4,6-dimethyl-2-oxo-2H-pyran-5-carboxylate 1 in alkaline medium, followed by decarboxylation, provides a simple route to the alpha, beta-unsaturated side-chain delta-lactone synthesis for a classical withanolide precursor.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service