Skip to Content
Merck
All Photos(1)

Key Documents

E21604

Sigma-Aldrich

Ethyl cyclopropanecarboxylate

99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€103.00

€103.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€103.00

About This Item

Linear Formula:
C3H5CO2C2H5
CAS Number:
Molecular Weight:
114.14
Beilstein:
2039752
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€103.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.420 (lit.)

bp

129-133 °C (lit.)

density

0.96 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C1CC1

InChI

1S/C6H10O2/c1-2-8-6(7)5-3-4-5/h5H,2-4H2,1H3

InChI key

LDDOSDVZPSGLFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haocheng Liu et al.
Foods (Basel, Switzerland), 9(1) (2020-01-16)
Mangoes (Mangifera indica L.) are wildly cultivated in China with different commercial varieties; however, characterization of their aromatic profiles is limited. To better understand the aromatic compounds in different mango fruits, the characteristic aromatic components of five Chinese mango varieties

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service