Skip to Content
Merck
All Photos(1)

Key Documents

E20705

Sigma-Aldrich

Ethyl cyclohexaneacetate

≥98%

Synonym(s):

Ethyl cyclohexylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH2CO2C2H5
CAS Number:
Molecular Weight:
170.25
Beilstein:
2043392
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
form:
liquid
Assay:
≥98%

Assay

≥98%

form

liquid

refractive index

n20/D 1.444 (lit.)

bp

211-212 °C/766 mmHg (lit.)

density

0.948 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC1CCCCC1

InChI

1S/C10H18O2/c1-2-12-10(11)8-9-6-4-3-5-7-9/h9H,2-8H2,1H3

InChI key

ZBDAMDWKXGTKBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service