Skip to Content
Merck
All Photos(2)

Key Documents

E12206

Sigma-Aldrich

2-Ethylanthraquinone

≥97%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€43.80

€43.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€43.80

About This Item

Empirical Formula (Hill Notation):
C16H12O2
CAS Number:
Molecular Weight:
236.27
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€43.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97%

mp

108-111 °C (lit.)

SMILES string

CCc1ccc2C(=O)c3ccccc3C(=O)c2c1

InChI

1S/C16H12O2/c1-2-10-7-8-13-14(9-10)16(18)12-6-4-3-5-11(12)15(13)17/h3-9H,2H2,1H3

InChI key

SJEBAWHUJDUKQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Blood,spleen

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service