DWKBGE820PNC
Azlon Dropper Bottles with spouted closure and cap
round bottom, low-density polyethylene, capacity 60 mL
Synonym(s):
BGE820PNC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121819
NACRES:
NB.43
Recommended Products
material
low-density polyethylene
round bottom
packaging
pkg of 10 ea
manufacturer/tradename
Azlon
capacity
60 mL
Looking for similar products? Visit Product Comparison Guide
Application
Suitable for as dropping bottle and excellent for dispensing gels
Features and Benefits
Pivotting nozzle closure
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service