Skip to Content
Merck
All Photos(1)

Key Documents

DWK92812F-10

KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask with PTFE stopper

glass flask, flask capacity (10 mL), class A

Synonym(s):

Heavy Duty Flask, Heavy Duty Volumetric Flask, Wide-Mouth Flask

Sign Into View Organizational & Contract Pricing

Select a Size

6 EA
€292.00

€292.00


Check Cart for Availability


Select a Size

Change View
6 EA
€292.00

About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

€292.00


Check Cart for Availability

material

PTFE stopper
glass flask

feature

class A

packaging

case of 6 ea

manufacturer/tradename

Kimble® 92812F-10

flask capacity

(10 mL)

Looking for similar products? Visit Product Comparison Guide

General description

Class A Heavy Duty Wide Mouth Volumetric Flasks with PTFE Stopper

  • Pipet access is easy with wide-mouth volumetric flasks
  • Superior strength, durability and safety from heavy, uniform walls
  • Quick identification with large, permanent, easy-to-read markings
  • Flat interior bottoms for stirring with standard stir bars
  • Graduated to Class A volumetric tolerances
  • Standard Taper glass stoppers
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements

Features and Benefits

  • Manufactured from borosilicate glass 3.3
  • Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
  • Heavy duty walls provide superior strength, durability and safety
  • Wide mouth minimizes glass-to-glass contact allowing greater flexibility in hand movements which reduces the risk of spills
  • Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
  • Printed large volume numbers allow quick identification when selecting glassware
  • Flat interior bottoms for stirring with standard stir bars
  • Calibrate to contain
  • Graduated to Class A volumetric tolerances

Legal Information

KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service