Key Documents
DWK73802-18400
KIMBLE® Solid Cap
18-400 thread, black phenolic cap (with rubber liner)
Select a Size
1 EACH
€178.00
€178.00
Check Cart for Availability
Select a Size
Change View
1 EACH
€178.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
€178.00
Check Cart for Availability
material
PTFE/rubber liner
black phenolic cap (with rubber liner)
sterility
non-sterile
feature
class A
packaging
case of 144 ea
manufacturer/tradename
Kimble® 73802-18400
fitting
18-400 thread
General description
Class A Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
  • Specially formulated phenolic cap material
  • Autoclavable
  • White rubber liners
  • Excellent for general laboratory use
Identified in ASTM Specification E982, Class A requirement
Features and Benefits
  • Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving
  • PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class A requirements
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)