Key Documents
DWK227497-00
Wheaton® Bod Bottle
capacity 300 mL, bottle diam. × H 43 mm × 115 mm, bottle diam. × H 69 mm × 165 mm, glass cap (Robotic Stopper)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
glass cap (Robotic Stopper)
Agency
suitable for SM 5210
packaging
pkg of 24 ea
manufacturer/tradename
WHEATON® 227497-00
bottle diam. × H
43 mm × 115 mm
69 mm × 165 mm
capacity
300 mL
General description
BOD Bottles are ideal for incubating diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during the stabilization of the decomposable organic matter by aerobic biochemical action. We recommend the 300mL capacity bottles for the five-day BOD test as referenced in Standard Methods for the Examination of Water and Wastewater, 21st Edition, 5210B, 2005.
Features and Benefits
Ideal for incubating diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during the stabilization of the decomposable organic matter by aerobic biochemical action
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Related Content
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.