Skip to Content
Merck
All Photos(1)

Documents

DWK211857307

Duran® Bottles, Reagent, Wide Neck

clear bottle (reagent)

Synonym(s):

211857307

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

clear bottle (reagent)
flat head glass cap (stopper)
wide-neck

Joint

Joints ST/NS neck (NS 85/55)

packaging

pkg of 1 ea

manufacturer/tradename

DURAN® 211857307

capacity

5000 mL

Looking for similar products? Visit Product Comparison Guide

General description

The classical stoppered reagent bottles are moulded from DURAN® borosilicate 3.3 Type 1 glass, and are suitable for the storage of liquid and solid chemicals and reagents. The bottles feature a wide conical ground joint neck. The wider neck is more convenient for handling powders. The glass stopper provides an excellent seal that protects the contents from evaporation, oxidation, and contamination. The bottles have a Retrace Code for manufacturing lot traceability and a marking field in permanent white enamel.

Application

Autoclaving of media and reagents, Preparation of media and reagents, Sample collection, Culturing vessel, Storage of media and reagents, Transport and shipping, Liquid handling reservoir, Boro 3.3

Features and Benefits

  • Manufactured from borosilicate glass 3.3
  • Easy access to the bottle contents due to the wide diameter of the bottle neck
  • DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
  • All borosilicate glass components and therefore suitable for storage of aggressive materials
  • Tight closure due to standard taper ground glass joint
  • Fired-on, highly durable white ceramic print

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service