Documents
DWK208923
Wheaton® Leak Resistant Bottle
capacity 8 mL, low-density polyethylene bottle, natural bottle, narrow-mouth bottle, bottle diam. × H 25 mm × 43 mm, 20-410
Synonym(s):
208923
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
low-density polyethylene bottle
narrow-mouth bottle
natural bottle
packaging
pkg of 72 ea
manufacturer/tradename
WHEATON® 208923
bottle diam. × H
25 mm × 43 mm
capacity
8 mL
Features and Benefits
Superior double seal closure for leak resistance
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.