Key Documents
DWK115300700-B
Wheaton® Microliter Vial
Select a Size
1 PKG
€300.00
€300.00
Check Cart for Availability
Select a Size
Change View
1 PKG
€300.00
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
€300.00
Check Cart for Availability
material
V-bottom polypropylene bottom
packaging
pkg of 1 packs
manufacturer/tradename
WHEATON® 11-5300-700-B
capacity
900 μL
Application
Chromatography
Features and Benefits
  • Double ring finish for added closure performance.
  • Approved for LC & GC.
  • Where noted, Certified Class 10,000 clean room packed enhances data integrity.
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)