Key Documents
DWK11-0051K
Wheaton® Microliter Snap Caps With Septa
black PTFE cap (Push fit)
Select a Size
100 EACH
€55.20
€55.20
Estimated to ship on19 April 2025
Select a Size
Change View
100 EACH
€55.20
About This Item
UNSPSC Code:
41100000
NACRES:
NB.11
€55.20
Estimated to ship on19 April 2025
material
black PTFE cap (Push fit)
feature
cap with septa
packaging
pkg of 100 ea
manufacturer/tradename
WHEATON® 11-0051K
Application
Chromatography
Features and Benefits
  • 11mm Snap Caps with installed septa offered in a wide variety of the most popular configurations.
  • Where noted, the Ring is installed between cap and septa for robotics employing magnetics to move vials.
  • Approved for GC and LC (where noted).
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)