Skip to Content
Merck
All Photos(1)

Key Documents

D91659

Sigma-Aldrich

Diethyl chlorophosphite

95%

Synonym(s):

Diethyl chlorophosphonite, Diethyl phosphorochloridite

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€49.40
25 G
€170.00
100 G
€523.00

€49.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€49.40
25 G
€170.00
100 G
€523.00

About This Item

Linear Formula:
(C2H5O)2PCl
CAS Number:
Molecular Weight:
156.55
Beilstein:
1098392
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€49.40


Check Cart for Availability

Request a Bulk Order

Assay

95%

form

liquid

refractive index

n20/D 1.434 (lit.)

bp

153-155 °C (lit.)
56-57.5 °C/30 mmHg (lit.)

density

1.089 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOP(Cl)OCC

InChI

1S/C4H10ClO2P/c1-3-6-8(5)7-4-2/h3-4H2,1-2H3

InChI key

TXHWYSOQHNMOOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethyl chlorophosphite can be used as a:
  • Reducing agent for the reduction of nitro compounds to the corresponding amines.[1]
  • Deoxygenating agent to reduce pyridine N-oxide to pyridine.[1]
  • Reagent in the conversion of aldoximes to nitriles via dehydration.[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

33.8 °F - closed cup

Flash Point(C)

1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service