Skip to Content
Merck
All Photos(1)

Documents

D68002

Sigma-Aldrich

4,5-Dichloro-2-nitroaniline

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(NO2)NH2
CAS Number:
Molecular Weight:
207.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

form

powder

mp

177-179 °C (lit.)

SMILES string

Nc1cc(Cl)c(Cl)cc1[N+]([O-])=O

InChI

1S/C6H4Cl2N2O2/c7-3-1-5(9)6(10(11)12)2-4(3)8/h1-2H,9H2

InChI key

FSGTULQLEVAYRS-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sławomir Kasperowicz et al.
IUBMB life, 72(6), 1211-1219 (2020-03-13)
A series of chlorine-substituted benzotriazole derivatives, representing all possible substitution patterns of halogen atoms attached to the benzotriazole benzene ring, were synthetized as potential inhibitors of human protein kinase CK2. Basic ADME parameters for the free solutes (hydrophobicity, electronic properties)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service