Skip to Content
Merck
All Photos(1)

Key Documents

D65607

Sigma-Aldrich

2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene

95%

Synonym(s):

2,4-Bis(chloromethyl)mesitylene

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€71.60

€71.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€71.60

About This Item

Linear Formula:
(CH3)3C6H(CH2Cl)2
CAS Number:
Molecular Weight:
217.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€71.60


Check Cart for Availability

Request a Bulk Order

Assay

95%

form

solid

bp

180 °C/20 mmHg (lit.)

mp

105-107 °C (lit.)

SMILES string

Cc1cc(C)c(CCl)c(C)c1CCl

InChI

1S/C11H14Cl2/c1-7-4-8(2)11(6-13)9(3)10(7)5-12/h4H,5-6H2,1-3H3

InChI key

JMNDJDCLOQRCJV-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service