Skip to Content
Merck
All Photos(1)

Key Documents

D49504

Sigma-Aldrich

Dibutyl sebacate

technical grade

Synonym(s):

Decanedioic acid dibutyl ester, Sebacic acid dibutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-(CH2)4CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
314.46
Beilstein:
1798308
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

grade

technical grade

form

liquid

refractive index

n20/D 1.441 (lit.)

bp

178-179 °C/3 mmHg (lit.)

solubility

hexane, toluene, ethanol and acetone: soluble
water and propylene glycol: insoluble

density

0.936 g/mL at 25 °C (lit.)

SMILES string

CCCCOC(=O)CCCCCCCCC(=O)OCCCC

InChI

1S/C18H34O4/c1-3-5-15-21-17(19)13-11-9-7-8-10-12-14-18(20)22-16-6-4-2/h3-16H2,1-2H3

InChI key

PYGXAGIECVVIOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibutyl sebacate(DBS) is a lipophilic dibutyl ester that can be used as a plasticizer with low solubility in water.[1][2]

Application

DBS can be used as a plasticizer for the formation of ethylcellulose based coating membranes which can be potentially used in drug delivery systems.[3] It may also be used as a plasticizer during the fabrication of electronic artificial tongue sensors for analyzing the intensity of olive oils.[4]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

366.8 °F - open cup

Flash Point(C)

186 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GC method for determining polyadipate plasticizers in foods: transesterification to dibutyl adipate, conversion to migrating polyadipate.
Biedermann M and Grob K
Chromatographia, 64(9-10), 543-552 (2006)
Physico-mechanical analysis of free ethylcellulose films plasticized with incremental weight percents of dibutyl sebacate.
Kangarlou S and Haririan I
Iranian Journal of Pharmaceutical Sciences, 3(3), 135-142 (2007)
Sensory intensity assessment of olive oils using an electronic tongue.
Veloso ACA, et al.
Talanta, 146(3), 585-593 (2016)
Sogol Kangarlou et al.
International journal of pharmaceutics, 356(1-2), 153-166 (2008-03-22)
This research was conducted to investigate the physico-mechanical characteristics of the EC-based coating membranes plasticized with two informal ingredients of vitamin resources, cholecalciferol and alpha-tocopherol, with respect to the commercial plasticizer DBS. Proceeding the experiment, free thin polymer sheetings of
A Wyss et al.
Biotechnology and bioengineering, 91(2), 227-236 (2005-05-26)
The activity of penicillin acylase has been studied in aqueous and organic solvents, as free enzyme as well as immobilized within the membrane of liquid-core capsules. The activity of the enzyme is inhibited by the accumulation of the products of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service